Learn More
Fisherbrand™ Polyurethane Chair - High Bench Height Medium Back, Chrome Foot Ring, and Stationary Glides in Polyurethane Products for Science Education
Fisherbrand™ Polyurethane Chairs with Medium Back are height adjustable for 42 in. work surface & feature ergonomic adjustments, chrome adjustable foot ring, & stationary glides. Self-skinning chemical, soil, & puncture resistant polyurethane makes this ideal for any environment.
$267.00 - $297.00
Specifications
Adjustable Yes
Material Polyurethane
Footrest Yes
With Armrest No
With Backrest Yes
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S40419 Products for Science Education
View Documents
Blue
Each for $267.00
Add to cart
S38664 Products for Science Education
View Documents
Black
Each for $297.00
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 23.25-33.25 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Chrome adjustable foot ring
  • Black, nylon-reinforced fiberglass base
  • Stationary, non-marring glides suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
Specifications
Specifications
Yes
Yes
Yes
No
Polyurethane
No
No