Learn More
Fisherbrand™ Polyurethane Chair - Medium Bench Height with Adjustable Arms, Chrome Foot Ring, and Casters in Black Polyurethane Products for Science Education
Fisherbrand™ Polyurethane Chairs are height adjustable for 36 in. work surface and feature ergonomic adjustments, arms, chrome adjustable foot ring, & self-braking casters. Self-skinning chemical, soil, and puncture resistant polyurethane makes this ideal for any environment.
$325.00 - $362.00
Specifications
Adjustable Yes
Material Polyurethane
Footrest Yes
With Armrest Yes
With Backrest Yes
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S38507 Products for Science Education
View Documents
Black
Each for $362.00
Add to cart
S40468 Products for Science Education
View Documents
Blue
Each for $325.00
Add to cart
Description
Description
  • Adjustable from 19.25-26.75 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Chrome adjustable foot ring
  • Black, nylon-reinforced fiberglass base
  • Dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
Specifications
Specifications
Yes
Yes
Yes
No
Polyurethane
Yes
Yes