Learn More
Fisherbrand™ Polyurethane Chair Round Tube Base - Desk Height with Adjustable Arms and Stationary Glides in Polyurethane Products for Science Education
Fisherbrand™ Polyurethane Chairs with Round Tube Base is height adjustable for 31 in. work surfaces & features ergonomic adjustments, larger back, adjustable arms, fixed foot ring & stationary glides. Self-skinning polyurethane is chemical, soil & puncture resistant & is easily cleaned.
$314.00 - $351.00
Specifications
Adjustable Yes
Material Polyurethane
Footrest No
With Armrest Yes
With Backrest Yes
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S36716 Products for Science Education
View Documents
Black
Each for $351.00
Add to cart
S40388 Products for Science Education
View Documents
Black
Each for $314.00
Add to cart
Description
Description
  • Adjustable from 19-24.25 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Stationary, non-marring glides suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
Specifications
Specifications
Yes
No
Yes
No
Polyurethane
Yes
No