Learn More
Fisherbrand™ Polyurethane Chair with Round Tube Base - Desk Height with Medium Back, Seat Tilt, Adjustable Arms, and Stationary Glides in Poly
Fisherbrand™ Polyurethane Chairs with Medium Back and Round Tube Base is height adjustable for 31 in. work surfaces and features ergonomic adjustments, larger back, adjustable arms, fixed foot ring and stationary glides. Self-skinning polyurethane is chemical, soil and puncture resistant and is easily cleaned.
$473.00 - $534.00
Specifications
Adjustable Yes
Base Material Powder-coated Steel
Material Polyurethane
Footrest No
With Armrest Yes
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
14-360-687
View Documents
Black
Each for $534.00
Add to cart
14-000-533
View Documents
Black
Each for $473.00
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 19-24.25 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 8-degree forward and 3-degree backward locking seat tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Stationary, non-marring glides suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
Specifications
Specifications
Yes
Polyurethane
Yes
No
Desk Height
Powder-coated Steel
No
Yes
Yes