Learn More
Fisherbrand™ Polyurethane Chair with Round Tube Base - Desk Height with Medium Back and Stationary Glides in Polyurethane
SDP
Fisherbrand™ Polyurethane Chairs with Medium Back and Round Tube Base is height adjustable for 31 in. work surfaces and features ergonomic adjustments, larger back, fixed foot ring, and stationary glides. Self-skinning polyurethane is chemical, soil, and puncture resistant and is easily cleaned.
$361.00 - $682.00
Specifications
Back Style Standard
Adjustable Yes
Base Style Powder-coated steel with fixed foot ring
Foot Type Black-non-marring bell glides (suitable for both hard and carpeted floors)
Footrest No
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
14-000-536
SDP
View Documents
Blue
Each for $361.00
Add to cart
14-360-443
SDP
View Documents
Black
Each for $682.00
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 19-24.25 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Stationary, non-marring glides suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
Specifications
Specifications
Standard
Powder-coated steel with fixed foot ring
No
Desk Height
No
No
19 to 24.25 in.
General Seating Chair
Yes
Black-non-marring bell glides (suitable for both hard and carpeted floors)
Polyurethane
32 lb.
Yes
No
Powder-coated Steel