Learn More
Fisherbrand™ Polyurethane Chair with Round Tube Base - High Bench Height with Medium Back and Stationary Glides in Polyurethane
Fisherbrand™ Polyurethane Chairs with Medium Back and Round Tube Base is height adjustable for 42 in. work surfaces and features ergonomic adjustments, larger back, fixed foot ring, and stationary glides. Self-skinning polyurethane is chemical, soil, and puncture resistant and is easily cleaned.
$372.00 - $718.50
Specifications
Adjustable Yes
Base Material Powder-coated Steel
Material Polyurethane
Footrest No
With Armrest No
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
14-000-572
View Documents
Black
Each for $372.00
Add to cart
14-360-449
View Documents
Black
Each for $718.50
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 26-36 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Stationary, non-marring glides suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
Specifications
Specifications
Yes
Polyurethane
No
No
High Bench Height
Powder-coated Steel
No
Yes
No