Learn More
Fisherbrand™ Polyurethane ESD Chair - High Bench Height with Chrome Foot Ring and Adjustable Arms Products for Science Education
Fisherbrand™ Polyurethane ESD Chairs are height adjustable for 42 in. work surfaces and feature ergonomic adjustments, electro-static dissipating components, adjustable arms, chrome adjustable foot ring, and self-skinning polyurethane. Ideal for static-sensitive applications.
$544.00 - $558.00
Specifications
Color Black
Adjustable Yes
Material Polyurethane
Footrest Yes
With Armrest Yes
View More Specs
Products 2
Catalog Number Foot Type Price Quantity & Availability  
Catalog Number Foot Type Price Quantity & Availability  
S38542 Products for Science Education
View Documents
Electro-Static Dissipating, dual-wheel, non-marring casters (suitable for both hard and carpeted floors)
Each for $558.00
Add to cart
S38470 Products for Science Education
View Documents
Electro-Static Dissipating, non-marring bell glides (suitable for both hard and carpeted floors)
Each for $544.00
Add to cart
Description
Description
  • Adjustable from 24.5-34.75 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Chrome adjustable foot ring
  • Electro-static dissipating components to ensure proper grounding
  • Cast-aluminum base
  • Black electro-static dissipating polyurethane
  • Meets ANSI/ESD STM12.1-2013
Specifications
Specifications
Black
Polyurethane
Yes
No
Yes
Yes
Yes
ANSI/ESD-STM-12.1-2006