Learn More
Fisherbrand™ Polyurethane Tractor Sit-Stand in Polyurethane
Fisherbrand™ Polyurethane Tractor Sit-Stand Stools are height adjustable for 42 in. work surfaces and features ergonomic adjustments, and stationary glides. Self-skinning polyurethane is chemical, soil, and puncture resistant, easily cleaned, and features breathable holes for added comfort.
Specifications
Adjustable Yes
Material Polyurethane
Footrest No
With Armrest No
With Backrest No
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
14-000-797
View Documents
Blue This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
14-000-796
View Documents
Black This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Adjustable from 22.5-32 inches from the top of the seat to the floor
  • Features ergonomic adjustments such as seat height
  • 8-degree forward and 3-degree backward locking seat tilt
  • Black, nylon-reinforced fiberglass base
  • Stationary, non-marring glides suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
Specifications
Specifications
Yes
No
No
No
Polyurethane
No
No
Stool