Learn More
Fisherbrand™ Polyurethane Tractor Sit-Stand Stool Chrome - Desk Height with Chrome Casters in Polyurethane
Fisherbrand™ Polyurethane Tractor Sit-Stand Stools in Chrome are height adjustable for 31 in. work surfaces & features ergonomic adjustments & self-braking casters. Self-skinning polyurethane is chemical, soil, & puncture resistant, easily cleaned, & features breathable holes for added comfort.
 View more versions of this product
Catalog No. 14-000-759
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Adjustable from 17.5-22.5 inches from the top of the seat to the floor
  • Features ergonomic adjustments such as seat height
  • Locking seat tilt
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
Specifications
Specifications
Yes
Chrome-hooded dual-wheel, self-braking, non-marring casters (suitable for both hard and carpeted floors)
Polyurethane
24 lb.
No
No
300 lb.
Tractor Sit/Stand Stool
Blue
No
Stool
No
Yes
17.5 to 22.5 in.
Polished Cast Aluminum
136 kg