Learn More
Fisherbrand™ Polyurethane Tractor Sit-Stand Stool - Desk Height with Casters in Polyurethane
SCI_ED
Fisherbrand™ Polyurethane Tractor Sit-Stand Stools are height adjustable for 31 in. work surfaces & features ergonomic adjustments & self-braking casters. Self-skinning polyurethane is chemical, soil, & puncture resistant, easily cleaned, & features breathable holes for added comfort.
 View more versions of this product
Catalog No. S41243
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Adjustable from 16.5-21.5 inches from the top of the seat to the floor
  • Features ergonomic adjustments such as seat height
  • Locking seat tilt
  • Black, nylon-reinforced fiberglass base
  • Dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
Specifications
Specifications
Yes
Black dual-wheel, self-braking, non-marring casters (suitable for both hard and carpeted floors)
Polyurethane
24 lb.
No
No
300 lb.
Tractor Sit/Stand Stool
Black
No
Stool
No
Yes
16.5 to 21.5 in.
Nylon Reinforced Fiberglass
136 kg