Learn More
Fisherbrand™ Polyurethane Tractor Stool Chrome - Medium Bench Height with Stationary Glides in Polyurethane Products for Science Education
Fisherbrand™ Polyurethane Tractor Stools in Chrome are height adjustable for 36 in. work surfaces & feature single-lever control mechanism & stationary glides. Self-skinning polyurethane is chemical, soil, & puncture resistant, easily cleaned, & features breathable holes for added comfort.
$132.00 - $141.00
Specifications
Adjustable Yes
Material Polyurethane
Footrest No
With Armrest No
With Backrest No
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S41266 Products for Science Education
View Documents
Blue
Each for $141.00
Add to cart
S41261 Products for Science Education
View Documents
Black
Each for $132.00
Add to cart
Description
Description
  • Adjustable from 20-27.5 inches from the top of the seat to the floor
  • Features ergonomic adjustments such as seat height
  • Cast-aluminum base
  • Stationary, non-marring glides suitable for hard floors
  • Chemical, soil, and puncture resistant polyurethane
Specifications
Specifications
Yes
No
No
No
Polyurethane
No
No
Stool