Learn More
Polyvinyl alcohol, 98-99% hydrolyzed, high molecular weight, Thermo Scientific Chemicals
Description
Polyvinyl alcohol is used in food industries as a binding and coating agent. Its viscosity favors the application as coating agent to tablets and capsules. Mixed-matrix membrane prepared by dispersion of NaA zeolite particles is used to study the pervaporative separation of water-butanol mixtures. Polyvinyl alcohol /gelatin based biocompatible polymeric scaffolds have been developed for 3D cancer models.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Solid |
Solubility Information | Soluble in water. |
Quantity | 25 g |
Percent Purity | 98 to 99% |
Odor | Odorless |
Chemical Name or Material | Polyvinyl alcohol, High molecular weight, Hydrolyzed |
Safety and Handling
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.