Learn More
Cytiva (Formerly GE Healthcare Life Sciences) Polyvinyl Chloride Fume Cupboard
This rich, robust, reliable, fume cupboard made of polyvinylchloride features glare fluorescent tube lightening.
$339.50
Specifications
Certifications/Compliance NFX 15-210 class B & EN 14175
Construction XPX 15-206
Lighting Glare fluorescent tube
Quantity 1Each
Type Fume Cupboard
View More Specs
Products 1
Catalog Number Mfr. No. Width (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Width (Metric) Price Quantity & Availability  
09-927-237
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
2642
150cm Pack of 5 for $339.50
Add to cart
Description
Description
  • Fume cupboard made of polyvinylchloride with glare fluorescent tube lightening
  • Standard sorbridge PVC (= registry fume cupboards)
  • PVC rear panel over the entire width of the fume hood
  • Guide the air at the top and bottom
  • Uses all types of solvents, volatile products, powders, condensing
Specifications
Specifications
NFX 15-210 class B & EN 14175
Glare fluorescent tube
Fume Cupboard
XPX 15-206
1Each
Polyvinylchloride