Alfa Aesar™ Polyvinylpyrrolidone-iodine complex

Manufacturer: Alfa Aesar™ 45782A1

Quantity 1kg

 View more versions of this product

Catalog No. AA45782A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 25655-41-8
    Molecular Formula (C6H9NO)n·xI
    MDL Number MFCD00084483
    Merck Index 14,7698
    Chemical Name or Material Polyvinylpyrrolidone-iodine complex

    Specifications

    Molecular Formula (C6H9NO)n·xI
    MDL Number MFCD00084483
    Merck Index 14,7698
    Quantity 1kg
    RTECS Number TR1579600
    TSCA TSCA
    Physical Form Powder
    CAS 25655-41-8
    Color Yellow-brown
    Melting Point 300°C (dec.)
    Chemical Name or Material Polyvinylpyrrolidone-iodine complex

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit