Alfa Aesar™ Polyvinylpyrrolidone, M.W. 1,300,000

Manufacturer: Alfa Aesar™ 4372818

Quantity 50g

 View more versions of this product

Catalog No. AA4372818

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 9003-39-8
    Molecular Formula (C6H9NO)n
    MDL Number MFCD00149016
    Merck Index 14,7697
    Chemical Name or Material Polyvinylpyrrolidone, M.W. 1,300,000

    Specifications

    TSCA TSCA
    Physical Form Powder
    Merck Index 14,7697
    Molecular Formula (C6H9NO)n
    CAS 9003-39-8
    Sensitivity Hygroscopic
    MDL Number MFCD00149016
    Odor Odorless
    Quantity 50g
    Chemical Name or Material Polyvinylpyrrolidone, M.W. 1,300,000

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit