Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POM121 Polyclonal Antibody
POM121 Polyclonal Antibody
Supplier: Thermo Scientific PA583979
Description
POM121 Polyclonal Antibody for ICC/IF, IHC (P)

Specifications
POM121 | |
Polyclonal | |
Unconjugated | |
POM121B | |
2610027A18Rik; C80273; KIAA0618; mKIAA0618; Nuclear envelope pore membrane protein POM 121; nuclear envelope pore membrane protein POM 121A; nuclear envelope pore membrane protein POM 121C; nuclear pore membrane glycoprotein 121 kD; nuclear pore membrane protein 121; nuclear pore membrane protein 121 kDa; nuclear pore membrane protein 121-2; nucleoporin Nup121; Nup121; p145; Pom121; POM121 membrane glycoprotein; POM121 membrane glycoprotein C; POM121 transmembrane nucleoporin; POM121 transmembrane nucleoporin C; POM121-2; POM121A; POM121B; POM121C; Pore membrane protein of 121 kDa; pore membrane protein of 121 kDa C | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100101267, 100288540, 9883 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NF01, A8CG34, Q96HA1 | |
POM121, POM121B, POM121C | |
Recombinant protein corresponding to Human POM121. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction