Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ POMP Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | POMP |
---|---|
Concentration | 500 μg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119365
|
Invitrogen™
PA5119365 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121673
|
Invitrogen™
PA5121673 |
20 μL |
Each for $209.50
|
|
|||||
Description
POMP Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Molecular chaperone essential for the assembly of standard proteasomes and immunoproteasomes. Degraded after completion of proteasome maturation. Mediates the association of 20S preproteasome with the endoplasmic reticulum.Specifications
POMP | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
POMP | |
2510048O06Rik; C13orf12; HSPC014; HSPC036; hUMP1; mUMP1; PNAS-110; POMP; Proteasome maturation protein; proteassemblin; Protein UMP1 homolog; RGD1305831; UMP1; Voltage-gated K channel beta subunit 4.1; voltage-gated potassium channel beta subunit 4.1 | |
POMP | |
Primary | |
Antigen affinity chromatography |
500 μg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q9Y244 | |
51371 | |
Recombinant protein Proteasome Maturation Protein. The antigen corresponds to amino acid range 1-141 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title