Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POMT1 Polyclonal Antibody
POMT1 Polyclonal Antibody
Supplier: Thermo Scientific PA542684
Description
POMT1 Polyclonal Antibody for Western Blot

Specifications
POMT1 | |
Polyclonal | |
Unconjugated | |
POMT1 | |
AI505244; dolichyl-phosphate-mannose--protein mannosyltransferase 1; FLJ37239; LGMD2K; LOW QUALITY PROTEIN: protein O-mannosyl-transferase 1; MDDGA1; MDDGB1; MDDGC1; POMT1; protein O-mannosyltransferase 1; Protein O-mannosyl-transferase 1; protein-O-mannosyltransferase 1; RT; testis tissue sperm-binding protein Li 57p | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10585 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y6A1 | |
POMT1 | |
Synthetic peptide directed towards the middle region of human POMT1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction