Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POMT1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | POMT1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP160054
|
Novus Biologicals
NBP160054 |
100 μL |
Each of 1 for $436.00
|
|
Description
POMT1 Polyclonal specifically detects POMT1 in Human samples. It is validated for Western Blot.Specifications
POMT1 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
Dolichyl-phosphate-mannose--protein mannosyltransferase 1, EC 2.4.1, EC 2.4.1.109, FLJ37239, LGMD2K, MDDGA1, MDDGB1, MDDGC1, protein O-mannosyl-transferase 1, protein-O-mannosyltransferase 1, RT | |
POMT1 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
B3KQG0 | |
10585 | |
Synthetic peptides corresponding to POMT1(protein-O-mannosyltransferase 1) The peptide sequence was selected from the middle region of POMT1. Peptide sequence LTFQILLLPVVLQHISDHLCRSQLQRSIFSALVVAWYSSACHVSNTLRPL. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title