Learn More
Invitrogen™ POMT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113831
Description
Immunogen sequence: RKHYQVTGYG INGTGDSNDF WRIEVVNRKF GNRIKVLRSR IRFIHLVTGC For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 85%.
The protein encoded by this gene is an O-mannosyltransferase that requires interaction with the product of the POMT1 gene for enzymatic function. The encoded protein is found in the membrane of the endoplasmic reticulum. Defects in this gene are a cause of Walker-Warburg syndrome (WWS).
Specifications
POMT2 | |
Polyclonal | |
Unconjugated | |
POMT2 | |
A830009D15Rik; AW046274; DKFZp686G10254; Dolichyl-phosphate-mannose--protein mannosyltransferase; dolichyl-phosphate-mannose--protein mannosyltransferase 2; FLJ22309; LGMD2N; MDDGA2; MDDGB2; MDDGC2; Pomt2; protein O-mannosyltransferase 2; Protein O-mannosyl-transferase 2; protein-O-mannosyltransferase 2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
29954 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UKY4 | |
POMT2 | |
Synthetic peptide directed towards the middle region of human POMT2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.