Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POMT2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | POMT2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP160068
|
Novus Biologicals
NBP160068 |
100 μL |
Each of 1 for $436.00
|
|
Description
POMT2 Polyclonal specifically detects POMT2 in Human samples. It is validated for Western Blot.Specifications
POMT2 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
29954 | |
Synthetic peptides corresponding to POMT2(protein-O-mannosyltransferase 2) The peptide sequence was selected from the middle region of POMT2. Peptide sequence AIGYLHSHRHLYPEGIGARQQQVTTYLHKDYNNLWIIKKHNTNSDPLDPS. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
DKFZp686G10254, Dolichyl-phosphate-mannose--protein mannosyltransferase 2, EC 2.4.1.109, FLJ22309, MDDGA2, MDDGB2, MDDGC2, protein O-mannosyl-transferase 2, protein-O-mannosyltransferase 2 | |
POMT2 | |
IgG | |
Affinity Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title