Learn More
PON1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 181551AP
Catalog No. 50-173-2020
Add to cart
Description
Description
Glycosylated PON1 migrates at 42-45 kDa. 55 kDa is a dimmer of isoform CRA-b. The antibody has cross-reaction to PON2 and PON3.
The enzyme encoded by this gene is an arylesterase that mainly hydrolyzes paroxon to produce p-nitrophenol. Paroxon is an organophosphorus anticholinesterase compound that is produced in vivo by oxidation of the insecticide parathion. Polymorphisms in this gene are a risk factor in coronary artery disease. The gene is found in a cluster of three related paraoxonase genes at 7q21.3.
Specifications
Specifications
PON1
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P27169, P52430, P55159, Q15165, Q15166, Q62086, Q62087, Q68FP2, Q6AXM8
A esterase 1, Aromatic esterase 1, ESA, K 45, paraoxonase 1, PON, PON 1, PON1, Serum aryldialkylphosphatase 1
Rabbit
IgG
150 μL
-20°C
Primary
18979, 269823, 296851, 312086, 330260, 5444, 5445, 5446, 84024
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
PON1
Liquid
Pon1, Pon2, PON3
PON1 Fusion Protein Ag12934
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat