Learn More
POP4 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 152731AP
Catalog No. 50-173-0109
Add to cart
Description
Description
Part of ribonuclease P, a protein complex that generates mature tRNA molecules by cleaving their 5 ft.-ends. May function with RPP38 to coordinate the nucleolar targeting and/or assembly of RNase P.
Specifications
Specifications
POP4
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O95707
hPOP4, POP4, RPP29
Rabbit
IgG
150 μL
-20°C
Primary
10775
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
POP4
Liquid
POP4
POP4 Fusion Protein Ag7387
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human