Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
POR/Cytochrome P450 Reductase Mouse anti-Human, Mouse, Rat, DyLight 650, Clone: OTI3F10, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273539C
Description
POR/Cytochrome P450 Reductase Monoclonal specifically detects POR/Cytochrome P450 Reductase in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
POR/Cytochrome P450 Reductase | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
POR | |
Full length recombinant protein of human POR/Cytochrome P450 Reductase (NP_000932) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI3F10 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
CPR, CYPORFLJ26468, DKFZp686G04235, EC 1.6.2.4, NADPH--cytochrome P450 reductase, NADPH-dependent cytochrome P450 reductase, P450 (cytochrome) oxidoreductase, P450R | |
Mouse | |
Affinity Purified | |
Breast Cancer, Stem Cell Markers | |
5447.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title