Learn More
United Scientific™ Porcelain Casserole
These casseroles offer lab grade porcelainware at substantial cost savings.
Supplier: United Scientific™ JPC060
This item is not returnable.
View return policy
Description
- Porcelain casserole, featuring lip and flat handle
- High resistance to acids and alkalis, not including hydrofluoric acid
- Autoclavable and glazed inside and out (except for rim and top of handle)
- Designed to withstand temperatures up to 1150°C
- To prevent thermal stress cracks, temperature change rate should no exceed 200°C per hour
Specifications
Porcelain | |
Autoclavable | |
33 mm |
60 mL | |
70 mm | |
Casserole |