Learn More
CoorsTek™ Porcelain Coal Moisture Testing Crucibles
Withstand temperatures up to 1150°C (2102°F), not to exceed 200°C/hour
Supplier:  CoorsTek™ 60052
 View more versions of this product
Catalog No. 08-103B
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Porcelain with rounded bottom
  • Glazed inside and out except on rim
  • 20mL size recommended for ASTM™ D 3176
  • Capsule form; without covers
Specifications
Specifications
Porcelain
25mL