Learn More
Novus Biologicals™ Porcine Atrial Natriuretic Peptide/ANP ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP266734
Catalog No. NBP266734
Add to cart
Description
Description
The Porcine Atrial Natriuretic Peptide/ANP ELISA Kit (Colorimetric) measures porcine Atrial Natriuretic Peptide/ANP in serum, plasma and other biological fluids.
Specifications
Specifications
1 Kit
15.63 - 1000 pg/mL
ELISA
ELISA Kit (Colorimetric)
Storage of components varies. See protocol for specific instructions.
Porcine
Biological Fluids, Plasma, Serum
This kit recognizes Porcine ANP in samples. No significant cross-reactivity or interference between Porcine ANP and analogues was observed.
4878
9.38 pg/mL
Atrial Natriuretic Peptide/ANP
NPPA