Learn More
Novus Biologicals™ Porcine C-Reactive Protein/CRP ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP267253
Description
The Porcine C-Reactive Protein/CRP ELISA Kit (Colorimetric) measures porcine C-Reactive Protein/CRP in serum, plasma and other biological fluids.Specifications
78.13 - 5000 pg/mL | |
This kit recognizes Porcine CRP in samples. No significant cross-reactivity or interference between Porcine CRP and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
1401 | |
Porcine | |
Storage of components varies. See protocol for specific instructions. |
46.88 pg/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
CRP | |
1 Kit | |
C-Reactive Protein/CRP |