Learn More
Raybiotech IncSupplier Diversity Partner Porcine Decorin ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELPDECORIN1
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 4 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA pair antibody detects Porcine Decorin and Human Decorin.
Catalog No. 50-195-0181