Learn More
Novus Biologicals™ Porcine Ferritin ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275371
Catalog No. NBP275371
Add to cart
Description
Description
The Porcine Ferritin ELISA Kit (Colorimetric) measures porcine Ferritin in serum, plasma and other biological fluids.
Specifications
Specifications
4.69 - 300 ng/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Porcine
1 Kit
This kit recognizes Porcine FE in samples. No significant cross-reactivity or interference between Porcine FE and analogues was observed.
2495
2.81 ng/mL
Ferritin
FTH1
Biological Fluids, Plasma, Serum