Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Porcine Free Tri-Iodothyronine ELISA Kit (Colorimetric)
Assay has high sensitivity and specificity for detection of pig FT3.
Supplier: Novus Biologicals™ NBP2600201KIT
Description
The Porcine Free Tri-Iodothyronine ELISA Kit (Colorimetric) measures porcine Free Tri-Iodothyronine in serum, plasma, cell culture supernates, tissue homogenates.Specifications
2pmol/L to 32pmol/L | |
ELISA Kit (Colorimetric) | |
For detection of pig FT3 | |
ELISA | |
1 Kit | |
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date. |
0.38pmol/L | |
Cell Culture Supernatant, Plasma, Serum, Tissue Homogenates | |
Porcine | |
Adhesive Strip (For 96 wells), Assay plate, Conjugate, HRP-avidin, Substrate A, Substrate B, Wash Buffer (20 x concentrate), Standard, Stop Solution, Instruction manual | |
50 to 100 μL | |
Free Tri-Iodothyronine |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction