Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Porcine IgG, F(ab), Fluorescein, Isotype control, Novus Biologicals™
Pig Antibody
Supplier: Novus Biologicals NBP196937
Description
IgG F(ab) specifically detects IgG F(ab) in Porcine samples. It is validated for Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.Specifications
IgG F(ab) | |
Fluorescein | |
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide | |
Multi-step | |
RUO | |
No reaction was observed against anti-Swine IgG F(c) or anti-Papain. | |
Porcine | |
IgG |
LYOPH | |
Restore with 1.0mL of deionized water (or equivalent). | |
Porcine | |
1 mg | |
Primary | |
Reconstitute with 1.0 ml deionized water (or equivalent). | |
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction