Learn More
Porcine Porcine IgG, Fc Protein, Novus Biologicals™
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP1970041MG
 View more versions of this product
Catalog No. NBP1970041
Add to cart
Description
Description
Native Porcine IgG, Fc. Source: Serum The Porcine IgG, Fc Native Protein has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, SDS-Page, Fluorophore-linked immunosorbent assay.
Specifications
Specifications
Enzyme Linked Immunosorbent Assay, Immunoprecipitation, SDS-Page, Western Blotting
Potassium Phosphate, 0.15M Sodium Chloride pH 7.2
1mg
Porcine IgG, Fc
Lyophilized
Ion exchange chromatography
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Unlabeled