Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Invitrogen™ Porcine IL-1 alpha/IL-1 F1 ELISA Kit Non-distribution product as customer accommodation.

Porcine IL-1 alpha/IL-1 F1 quantitates porcine IL-1 alpha/IL-1 F1 in serum, plasma, supernatant.

Specifications

Regulatory Status Research Use Only
Storage Requirements 2°C to 8°C
Target IL-1 alpha
Product Type ELISA
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ES10RBX5 Non-distribution product as customer accommodation.
View Documents
Invitrogen™
ES10RBX5
5 x 96 Tests
Each for $2,205.00
N/A
ES10RB Non-distribution product as customer accommodation.
View Documents
Invitrogen™
ES10RB
96 Tests
Each for $491.00
N/A
Description

Description

Porcine IL-1 alpha/IL-1 F1 quantitates porcine IL-1 alpha/IL-1 F1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant porcine IL-1 alpha/IL-1 F1.

IL-1 alpha (Interleukin-1 alpha) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 alpha is coded by the IL1A gene and signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 beta. IL1A is located on the q arm on chromosome 2 at position 13. IL-1 alpha plays an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses. IL-1 alpha activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 alpha is a pleiotropic cytokine involved in various immune responses, inflammatory processes, and hematopoiesis. Further, IL-1 alpha is produced by monocytes and macrophages as a proprotein, which is proteolytically processed and released in response to cell injury and induces apoptosis. The IL1A gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. IL1 alpha regulates the activities of NF-kappaB and mitogen-activated protein kinases. IL-1 alpha can stimulate the expression of IL6 and cyclooxygenase-2 (PTGS2/COX-2), as well as enhance the production of nitric oxide (NO). High levels of IL-1 alpha are associated with several chronic inflammatory diseases including rheumatoid arthritis, psoriasis and multiple sclerosis.
Specifications

Specifications

Research Use Only
IL-1 alpha
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
4 hr 45 min
Serum, Plasma, Supernatant
2°C to 8°C
ELISA
4.5 pg/mL
1 hr 20 min
Plasma, 50 µL; Supernatant, 50 µL; Serum, 50 µL
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit