Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Porcine IL-4 ELISA Kit Non-distribution product as customer accommodation.

Porcine IL-4 quantitates porcine IL-4 in serum, plasma, supernatant.

$491.00

Specifications

Storage Requirements 2°C to 8°C
Assay Sensitivity 1.5 pg/mL
Assay Range 1.5 to 400 pg/mL
Test Time 1 hr 20 min
Sample Volume Plasma, 50 µL; Supernatant, 50 µL; Serum, 50 µL
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ES14RB Non-distribution product as customer accommodation.
View Documents
Invitrogen™
ES14RB
96 Tests
Each for $491.00
Add to cart
 
Description

Description

Porcine IL-4 quantitates porcine IL-4 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant porcine IL-4.

Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.
Specifications

Specifications

2°C to 8°C
1.5 to 400 pg/mL
Plasma, 50 µL; Supernatant, 50 µL; Serum, 50 µL
<10%
1.5 pg/mL
1 hr 20 min
<12%
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit