Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Porcine Rotavirus Antigen ELISA Kit (Colorimetric)

Assay has high sensitivity and specificity for detection of pig Rotavirus antigen

$602.00

Specifications

Species Porcine
Sample Type feces
Research Discipline Cell Biology
Specificity For detection of pig Rotavirus antigen
Storage Requirements Store the unopened product at 2 to 8°C Protect from light. Do not use past expiration date.
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB601331KT View Documents Novus Biologicals™
NBP2601331KIT
1 Kit Each for $602.00
Add to cart
 
Description

Description

The Porcine Rotavirus Antigen ELISA Kit (Colorimetric) measures Porcine Rotavirus Antigen in feces.
Specifications

Specifications

Porcine
Cell Biology
Store the unopened product at 2 to 8°C Protect from light. Do not use past expiration date.
ELISA
Qualitative
Adhesive Strip (For 96 wells), Assay plate, HRP-conjugate, Substrate B, Substrate A, Wash Buffer, Stop Solution, Negative Control, Positive Control, Instruction manual
feces
For detection of pig Rotavirus antigen
Rotavirus Antigen
ELISA Kit (Colorimetric)
50 - 100μL

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit