Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Porcine Triiodothyronine ELISA Kit (Colorimetric)

Assay has high sensitivity and excellent specificity for detection of pig T3

$602.00

Specifications

Species Porcine
Sensitivity 500pg/mL
Sample Type serum, plasma
Research Discipline Signal Transduction
Specificity For detection of pig T3
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB601841KT View Documents Novus Biologicals™
NBP2601841KIT
1 Kit Each for $602.00
Add to cart
 
Description

Description

The Porcine Triiodothyronine ELISA Kit (Colorimetric) measures porcine Triiodothyronine in serum, plasma.
Specifications

Specifications

Porcine
serum, plasma
For detection of pig T3
Store the unopened product at 2 to 8°C Protect from light. Do not use past expiration date.
ELISA
500 pg/ml to 8000 pg/ml
Adhesive Strip (For 96 wells), Assay plate, Conjugate, HRP-avidin, Wash Buffer (20 x concentrate), Substrate B, Substrate A, Standard, Stop Solution, Instruction manual
500pg/mL
Signal Transduction
liothyronine; T3
Triiodothyronine
ELISA Kit (Colorimetric)
50 - 100μL

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit