Learn More
Porifera Sponges Products for Science Education
Become familiar with the least-developed organism of the animal kingdom.
$16.77
Specifications
Packaging Jar
View More Specs
Products 5
Catalog Number Length (English) Preservative Product Type Quantity Price Quantity & Availability  
Catalog Number Length (English) Preservative Product Type Quantity Price Quantity & Availability  
S1002 Products for Science Education
View Documents
0.5 to 0.75 in. Alcohol Grantia Sponge 10 Pack
Pack of 10 for $17.98
N/A
Due to product restrictions, please to purchase or view availability for this product.
S1015S Products for Science Education
View Documents
0.25 to 0.5 in. Alcohol Grantia Sponge 10 Pack
Pack of 10 for $15.79
N/A
Due to product restrictions, please to purchase or view availability for this product.
S65074 Products for Science Education
View Documents
1 to 2 in. Formaldehyde Sponge survey set Each
Each for $16.77
Add to cart
S1221S Products for Science Education
View Documents
5 to 6 in. Formaldehyde Commercial Sponge 10 Pack This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
S1210S Products for Science Education
View Documents
1 to 2 in. Alcohol Fresh Water Sponge Ea. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
Description
Description
Grantia, 1/4 to 3/4 in. (Scypha)
  • A small sponge characterized by its vase or urn shape
  • Has one large excurrent pore called an osculum, located at the upper end of the body
  • Larger sizes have become scarce recently; however, smaller-size specimens are readily available
  • In alcohol
Commercial sponge, 5 to 6 in.
  • Portions of this dry sponge provide an excellent “hands-on” specimen
  • Many pores penetrate the sponge and are readily apparent
  • Dry
Freshwater sponge
  • Spongilla, a Rhagon-type sponge that is a common inhabitant of clean ponds, lakes, streams and rivers
  • In alcohol
Complete Study Set
  • Set contains one each of Grantia, Spongilla and a commercial sponge
  • Each sponge comes in a labeled jar
Specifications
Specifications
Jar