Learn More
Echelon Biosciences Research LabsSupplier Diversity Partner PORPHOBILINOGEN 10 mg  DFS Item
Manufacturer:  Echelon Biosciences Research LabsSupplier Diversity Partner FH02010MG
Porphobilinogen is an intermediate in the heme synthesis pathway produced from d-Amino-levulinic acid in a reaction catalyzed by the enzyme ALA dehydrogenase. Porphobilinogen is converted to Hydroxylmethyl bilane in a reaction catalyzed by the enzyme Porphobilinogen Deaminase.
Catalog No. 50-720-329