Learn More
Fisherbrand™ Port Disk with 1/4 in. Barb - E8402 - QC
Superior single barb technology that provides a leak-proof seal. Ideal with flexible tubing. Designed for Sample Port and Dip Tube Applications.
Description
- Single & Multi Use Applications
- PVC-Free
- Superior Single Barb Design
- USP Class VI Compliant
- RoHS Compliant
- Gamma Stable
- Animal Derivative Free
- Lot Traceable
Specifications
Class 7 Cleanroom Manufactured | |
Bag Port | |
Biomedical and laboratory environment in fluid management applications. |