Learn More
Fisherbrand™ Port Disk with 3/16 in. Barb - E8402 - QC
SDP
Superior single barb technology that provides a leak-proof seal. Ideal with flexible tubing.
Catalog No. 01-000-094
Add to cart
Description
Description
  • Mini-Dome protrusions inhibit suction blockage and has a flexible 2 ¼” disc base.
  • Bag ports, also known as port plates and port discs, are designed to be ultrasonic or thermal welded to medical and bio-reactor bags of like material.
  • These single-use bags are used to transfer drug compounds, cell culture and fermentation as well as other media and fluids in the bio-pharmaceutical industry.
  • Single & Multi Use Applications
  • PVC-Free
  • Superior Single Barb Design
  • USP Class VI Compliant
  • RoHS Compliant
  • Gamma Stable
  • Animal Derivative Free
  • Lot Traceable
Warranty and Services
Replacement if not satisfied. 12 Month return Policy.
Specifications
Specifications
Compliance Certificates Available
Engage E8402
Single-use pharmaceutical bio-reactor bag applications. Biomedical and laboratory environment in fluid management applications. 
Out of direct sunlight, >20°F <100°F
Class 7 Cleanroom Manufactured
Bag Port
100 Pack
Natural