Learn More
HANNA INSTRUMENTS INC Free and Total Chlorine Portable Photometer  DFS Item
Manufacturer:  HANNA INSTRUMENTS INC HI96711C
The HI96711 Free and Total Chlorine Portable Photometer combines accuracy and ease of use in an ergonomic, portable design. A user can accurately determine the concentration of free and total chlorine in a variety of water samples within a 0.00 to 5.00 mg/L (ppm) range using ready made reagents. The HI93701-0 reagents are used for free chlorine and the the HI93711-0 are used for total chlorine measurements. The HI96711 offers many advanced features including the exclusive CAL Check™ function used for performance verification and calibration of the meter.CAL Check™ Validation and CalibrationBuilt-in TimerAutomatic Shut Off
Catalog No. NC1722447
Please call customer service for current pricing. 1-800-766-7000