Learn More
Brady™ Portable Metal Lock Box
Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Portable Metal Lock Box is made of heavy-duty steel and is powder coated for extra rust resistance. Lock boxes require fewer locks and are lighter than hasps on delicate energry isolating devices like circuit breakers.
Manufacturer:  Brady™ 73289
Catalog No. 19-104-095
Add to cart
Description
Description
  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
Safety
Portable
6 in.
Powder-coated steel
3.5 in. x 9 in. x 6 in.
1/Lock box
Red
3.5 in.
9 in.