Learn More
SI Instruments Portable Rubber Durometer
Supplier:  SI Instruments 70300/DEL
Catalog No. 14-202
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.
Description
Description
For hardness testing of plastics and cellular, soft, and hard rubbers. Scale graduations 0 to 100 in one-point increments. Place durometer in contact with material. Apply pressure sufficient to ensure firm contact between durometer flat bottom and sample. Read gauge immediately. Application pressure should not exceed 1kg.Includes protective vinyl-covered steel case and test block for checking accuracy. Without support stand.
Compliance
Meets specifications of ASTM D 2240-00.
Product Suggestions
Product Suggestions