Learn More
Portable Steam Sterilizer: Support Base Products for Science Education
Optional support base for use with Portable Steam Sterilizer (S50394M). Elevates electric sterilizer above table or counter surface to prevent heat damage.
Manufacturer:  Wisconsin Aluminum Foundry Company Inc 2180/EMD
Catalog No. S50396
Add to cart
Specifications
Specifications
Education
12.37 in.
12.75 in.
Portable Steam Sterilizer
5cm
17 to 19 psi
UL
31.4cm
32.4cm
2 in.
Support Base for Sterilizer