Alfa Aesar™ Potassium 2,4-pentanedionate hydrate, 97%

Manufacturer: Alfa Aesar™ B2416514

Quantity 25g

 View more versions of this product

Catalog No. AAB2416514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 57402-46-7
    Molecular Formula C5H7KO2·xH2O
    Formula Weight 138.21 (Anhydrous)
    MDL Number MFCD00150167
    Synonym Potassium acetylacetonate hemihydrate
    Chemical Name or Material Potassium 2,4-pentanedionate hydrate

    Specifications

    Formula Weight 138.21 (Anhydrous)
    Molecular Formula C5H7KO2·xH2O
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point ≈232°C (dec.)
    MDL Number MFCD00150167
    CAS 57402-46-7
    Synonym Potassium acetylacetonate hemihydrate
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Quantity 25g
    Chemical Name or Material Potassium 2,4-pentanedionate hydrate
    Assay 97%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit