Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Potassium 2-(azidomethyl)phenyltrifluoroborate, 95%, Thermo Scientific™

Click to view available options
Quantity:
1g
898544-48-4
C7H6BF3KN3
239.05
MFCD11052751
AKJNDNIFSIXTBR-UHFFFAOYSA-N
potassium 2-azidomethyl phenyltrifluoroborate, potassium 2-azidomethyl phenyl trifluoroborate, potassium 2-azidomethyl phenyl trifluoroboranuide, trifluoro 2-azidomethyl phenyl potassioboron v, potassium 2-azidomethyl phenyl trifluoro borate 1-
57416898
potassium;[2-(azidomethyl)phenyl]-trifluoroboranuide
[B-](C1=CC=CC=C1CN=[N+]=[N-])(F)(F)F.[K+]
898544-48-4
C7H6BF3KN3
239.05
MFCD11052751
AKJNDNIFSIXTBR-UHFFFAOYSA-N
potassium 2-azidomethyl phenyltrifluoroborate, potassium 2-azidomethyl phenyl trifluoroborate, potassium 2-azidomethyl phenyl trifluoroboranuide, trifluoro 2-azidomethyl phenyl potassioboron v, potassium 2-azidomethyl phenyl trifluoro borate 1-
57416898
potassium;[2-(azidomethyl)phenyl]-trifluoroboranuide
[B-](C1=CC=CC=C1CN=[N+]=[N-])(F)(F)F.[K+]
Quantity 1g
Formula Weight 241.06
Percent Purity 95%
Chemical Name or Material Potassium 2-(azidomethyl)phenyltrifluoroborate

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures