Alfa Aesar™ Potassium 2-ethylhexanoate, 99.9% (metals basis), 75% w/w soln.

Manufacturer: Alfa Aesar™ 3916309

Quantity 10g

 View more versions of this product

Catalog No. AA3916309

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3164-85-0
    Linear Formula K[OOCCH(C2H5)C4H9]
    Formula Weight 182.3
    MDL Number MFCD00045896
    UN Number UN1993
    Chemical Name or Material Potassium 2-ethylhexanoate

    Specifications

    Physical Form Viscous Liquid
    Formula Weight 182.3
    Odor Characteristic
    Health Hazard 3 GHS P Statement
    P280h-P273
    Wear protective gloves/clothing.
    Avoid release to the environment.
    MDL Number MFCD00045896
    Health Hazard 1 Warning
    Quantity 10g
    Linear Formula K[OOCCH(C2H5)C4H9]
    UN Number UN1993
    TSCA TSCA
    CAS 3164-85-0
    Health Hazard 2 GHS H Statement
    H226-H361-H373
    Flammable liquid and vapor.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Concentration 75% w/w soln.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Chemical Name or Material Potassium 2-ethylhexanoate
    Assay 99.9%, (metals basis)

    GHS H Statement
    H226-H361-H373
    Flammable liquid and vapor.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P280h-P273
    Wear protective gloves/clothing.
    Avoid release to the environment.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit