Alfa Aesar™ Potassium 4-acetylphenyltrifluoroborate, 97%

Manufacturer: Alfa Aesar™ H5584503

Quantity 1g

 View more versions of this product

Catalog No. AAH5584503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 252726-24-2
    Formula Weight 226.05
    MDL Number MFCD07784376
    Chemical Name or Material Potassium 4-acetylphenyltrifluoroborate

    Specifications

    Formula Weight 226.05
    MDL Number MFCD07784376
    Color White to off-white
    Melting Point 290°C (dec.)
    Physical Form Powder
    CAS 252726-24-2
    Quantity 1g
    Chemical Name or Material Potassium 4-acetylphenyltrifluoroborate
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit