Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium 4-(azidomethyl)phenyltrifluoroborate, 95%, Thermo Scientific™

Click to view available options
Quantity:
1g
5g
898544-50-8
C7H6BF3KN3
239.05
MFCD22124731
XIWGKBHBLASHCL-UHFFFAOYSA-N
potassium 4-azidomethyl phenyltrifluoroborate, potassium 4-azidomethyl phenyl trifluoroborate, potassium 4-azidomethyl phenyl trifluoroboranuide, c7h6bkn3, amtb514, trifluoro 4-azidomethyl phenyl potassioboron v
71306586
potassium;[4-(azidomethyl)phenyl]-trifluoroboranuide
[B-](C1=CC=C(C=C1)CN=[N+]=[N-])(F)(F)F.[K+]
898544-50-8
C7H6BF3KN3
239.05
MFCD22124731
XIWGKBHBLASHCL-UHFFFAOYSA-N
potassium 4-azidomethyl phenyltrifluoroborate, potassium 4-azidomethyl phenyl trifluoroborate, potassium 4-azidomethyl phenyl trifluoroboranuide, c7h6bkn3, amtb514, trifluoro 4-azidomethyl phenyl potassioboron v
71306586
potassium;[4-(azidomethyl)phenyl]-trifluoroboranuide
[B-](C1=CC=C(C=C1)CN=[N+]=[N-])(F)(F)F.[K+]
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Quantity 5g
Formula Weight 241.06
Percent Purity 95%
Chemical Name or Material Potassium 4-(azidomethyl)phenyltrifluoroborate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures